Ethyl (4-nitrobenzoyl)acetate ≥98.0%

Supplier: TCI America

Synonyms: 3-(4-Nitrophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Nitrobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Nitrophenyl)-3-oxopropionate

N0513-5G N0513-25G
TCN0513-5GEA 69.52 CAD
TCN0513-5G TCN0513-025G
Ethyl (4-nitrobenzoyl)acetate ≥98.0%
Ethyl (4-nitrobenzoyl)acetate

CAS Number: 838-57-3
MDL Number: MFCD00007357
Molecular Formula: C11H11NO5
Molecular Weight: 237.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: (4-Nitrobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Nitrophenyl)-3-oxopropionate, 3-(4-Nitrophenyl)-3-oxopropionic Acid Ethyl Ester

Formula: C₁₁H₁₁NO₅
MW: 237.21 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00007357
CAS Number: 838-57-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=186190120a8b65685ca923ad3641edd2e2d349a8&vid=186190120a8b65685ca923ad3641edd2e2d349a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR