4-Nitrostyrene ≥98.0% stabilized
Supplier: TCI America
N0503-1G
N0503-5G
TCN0503-001GEA
154.29
CAD
TCN0503-001G
TCN0503-005G
4-Nitrostyrene ≥98.0% stabilized
4-Nitrostyrene
(stabilized with TBC)
CAS Number: 100-13-0
MDL Number: MFCD00041254
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 109
Freezing point (°C): 21
Storage Temperature: <0°C
Formula:
C₈H₇NO₂ Flash Pt: 109 °C |
MDL Number:
MFCD00041254 CAS Number: 100-13-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...