Neopentyl glycol mono(hydroxypivalate) ≥97.0%

Supplier: TCI America

Synonyms: 2,2-Dimethyl-1,3-propanediol Mono(hydroxypivalate), 3-Hydroxy-2,2-dimethylpropyl 3-Hydroxy-2,2-dimethylpropionate, Hydroxypivalic Acid Neopentyl Glycol Ester

N0461-25G N0461-500G
TCN0461-025GEA 53.72 CAD
TCN0461-025G TCN0461-500G
Neopentyl glycol mono(hydroxypivalate) ≥97.0%
Neopentyl glycol mono(hydroxypivalate)

CAS Number: 1115-20-4
MDL Number: MFCD00059597
Molecular Formula: C10H20O4
Molecular Weight: 204.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 292
Melting point (°C): 51
Flash Point (°C): 161


Other Product Names / Synonyms: 2,2-Dimethyl-1,3-propanediol Mono(hydroxypivalate), Hydroxypivalic Acid Neopentyl Glycol Ester, 3-Hydroxy-2,2-dimethylpropyl 3-Hydroxy-2,2-dimethylpropionate

Formula: C₁₀H₂₀O₄
Boiling Pt: 292 °C
Melting Pt: 51 °C
Flash Pt: 161 °C
MDL Number: MFCD00059597
CAS Number: 1115-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=402ef1d46a65e669e95217b065cd8903d6dedd18&vid=402ef1d46a65e669e95217b065cd8903d6dedd18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR