4-Nitrobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0435-5G N0435-25G
TCN0435-005GEA 790.54 CAD
TCN0435-005G TCN0435-025G
4-Nitrobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzylamine hydrochloride

CAS Number: 18600-42-5
MDL Number: MFCD00012863
Molecular Formula: C7H8N2O2
Molecular Weight: 188.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 265

Formula: C₇H₉ClN₂O₂
Melting Pt: 265 °C
MDL Number: MFCD00012863
CAS Number: 18600-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e7207a97d72e1d502a64185321cb105804db8f&vid=a7e7207a97d72e1d502a64185321cb105804db8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR