Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97.0% (by HPLC)
Supplier: TCI America
N0369-25G
N0369-250G
TCN0369-025GEA
203.35
CAD
TCN0369-025G
TCN0369-250G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97.0% (by HPLC)
Naphthalene-1,4,5,8-tetracarboxylic dianhydride
CAS Number: 81-30-1
MDL Number: MFCD00006915
Molecular Formula: C14H4O6
Molecular Weight: 268.18
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Organic Transistor Materials, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: NTCDA
Formula:
C₁₄H₄O₆ |
MDL Number:
MFCD00006915 CAS Number: 81-30-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...