Dimethyl-2,6-naphthalenedicarboxylate ≥99.0%

Supplier: TCI America

Synonyms: Dimethyl 2,6-naphthalenedicarboxylate, 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester

N0364-25G N0364-5G
TCN0364-025GEA 135.88 CAD
TCN0364-025G TCN0364-5G
Dimethyl-2,6-naphthalenedicarboxylate ≥99.0%
Dimethyl-2,6-naphthalenedicarboxylate

CAS Number: 840-65-3
MDL Number: MFCD00004100
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 192
Flash Point (°C): 202


Other Product Names / Synonyms: 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
Melting Pt: 192 °C
Flash Pt: 202 °C
MDL Number: MFCD00004100
CAS Number: 840-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b8b181464b16e4302c8190a58035a3e49e1e152&vid=7b8b181464b16e4302c8190a58035a3e49e1e152&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR