5-Norbornene-2,3-dicarboxylic anhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

N0331-25G N0331-500G
TCN0331-025GEA 50.48 CAD
TCN0331-025G TCN0331-500G
5-Norbornene-2,3-dicarboxylic anhydride ≥97.0% (by titrimetric analysis)
5-Norbornene-2,3-dicarboxylic anhydride

CAS Number: 826-62-0
MDL Number: MFCD00069013
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 165


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Anhydride

Formula: C₉H₈O₃
Melting Pt: 165 °C
MDL Number: MFCD00069013
CAS Number: 826-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01447a6e71fbfdf3fac41ebfb939b02887a7905d&vid=01447a6e71fbfdf3fac41ebfb939b02887a7905d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR