2-Mercapto-6-nitrobenzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 6-Nitrobenzo[d]thiazole-2(3H)-thione

N0326-25G N0326-5G
TCN0326-025GEA 709.1 CAD
TCN0326-025G TCN0326-005G
2-Mercapto-6-nitrobenzothiazole ≥98.0%
2-Mercapto-6-nitrobenzothiazole

CAS Number: 4845-58-3
MDL Number: MFCD00041850
Molecular Formula: C7H4N2O2S2
Molecular Weight: 212.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Nitro-2-mercaptobenzothiazole

Formula: C₇H₄N₂O₂S₂
MDL Number: MFCD00041850
CAS Number: 4845-58-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR