1-Nonanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium 1-nonanesulfonate, Sodium nonane-1-sulfonate, Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

N0311-25G N0311-5G
TCN0311-025GEA 186.76 CAD
TCN0311-025G TCN0311-5G
1-Nonanesulfonic acid sodium salt ≥98.0%
1-Nonanesulphonic acid sodium salt

CAS Number: 35192-74-6
MDL Number: MFCD00025010
Molecular Formula: C9H20O3S
Molecular Weight: 230.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Nonylsulfonic Acid Sodium Salt, 1-Nonanesulfonic Acid Sodium Salt

Formula: C₉H₁₉NaO₃S
MDL Number: MFCD00025010
CAS Number: 35192-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c3688550962c5bddd0a4ef84fdc8335afe17e18&vid=7c3688550962c5bddd0a4ef84fdc8335afe17e18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR