Acid green 1 ≥80.0% (by UV method)

Supplier: TCI America

N0306-500G N0306-25G
TCN0306-500GEA 712.58 CAD
TCN0306-500G TCN0306-025G
Acid green 1 ≥80.0% (by UV method)
Acid green 1

CAS Number: 19381-50-1
MDL Number: MFCD00003886
Molecular Formula: C30H18FeN3O15S3
Molecular Weight: 878.45
Purity/Analysis Method: >80.0% (E)
Form: Crystal
Lambda max.: 714 nm (0.02mol/L AcONH4 sol.)


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Naphthol Green B

Formula: C₃₀H₁₈FeN₃O₁₅S₃
MDL Number: MFCD00003886
CAS Number: 19381-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7895e851f24b8c38e733b3289274a025b67351a3&vid=7895e851f24b8c38e733b3289274a025b67351a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR