1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt ≥98.0% (by titrimetric analysis)
Supplier: TCI America
N0268-25G
N0268-500G
TCN0268-025GEA
98.72
CAD
TCN0268-025G
TCN0268-500G
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt ≥98.0% (by titrimetric analysis)
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt
CAS Number: 525-05-3
MDL Number: MFCD00003885
Molecular Formula: C10H7NO8S2
Molecular Weight: 377.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Disodium 3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonate, 3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonic Acid Disodium Salt, 1-Nitroso-2-naphthol-3,6-disulfonic Acid Disodium Salt, Nitroso R Salt
Formula:
C₁₀H₅NNa₂O₈S₂ |
MDL Number:
MFCD00003885 CAS Number: 525-05-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...