4-(4-Nitrophenylazo)resorcinol

Supplier: TCI America

Synonyms: 4-((4-Nitrophenyl)diazenyl)benzene-1,3-diol, Azoviolet, Azo Violet, Magneson I

N0135-1G N0135-25G
TCN0135-001GEA 30.56 CAD
TCN0135-001G TCN0135-025G
4-(4-Nitrophenylazo)resorcinol
4-(4-Nitrophenylazo)resorcinol

CAS Number: 74-39-5
MDL Number: MFCD00007310
Molecular Formula: C12H9N3O4
Molecular Weight: 259.22
Form: Crystal
Color: Yellow Red
Melting point (°C): 192


Other Product Names / Synonyms: Azo Violet, 4-(4-Nitrophenylazo)resorcinol

Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
Melting Pt: 192 °C
MDL Number: MFCD00007310
CAS Number: 74-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86415db6233cf982e68e81e3f29c598976d911e6&vid=86415db6233cf982e68e81e3f29c598976d911e6&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR