Sodium nicotinate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0089-25G N0089-500G
TCN0089-025GEA 53.72 CAD
TCN0089-025G TCN0089-500G
Sodium nicotinate ≥98.0% (by titrimetric analysis)
Sodium nicotinate

CAS Number: 54-86-4
MDL Number: MFCD00064352
Molecular Formula: C6H5NO2
Molecular Weight: 145.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Pyridine-3-carboxylic Acid Sodium Salt, Nicotinic Acid Sodium Salt, Sodium Pyridine-3-carboxylate

Formula: C₆H₄NNaO₂
MW: 145.10 g/mol
MDL Number: MFCD00064352
CAS Number: 54-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=531f1c82756a826b4dd6c347b8f626baa7b09ec0&vid=531f1c82756a826b4dd6c347b8f626baa7b09ec0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR