α-Naphtholphthalein ≥98.0%

Supplier: TCI America

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

N0031-1G N0031-5G
TCN0031-001GEA 53.72 CAD
TCN0031-001G TCN0031-5G
α-Naphtholphthalein ≥98.0%
α-Naphtholphthalein

CAS Number: 596-01-0
MDL Number: MFCD00036202
Molecular Formula: C28H18O4
Molecular Weight: 418.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 240

Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00036202
CAS Number: 596-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24b7f5408fb35e318f3dd203f02eaca27daeb293&vid=24b7f5408fb35e318f3dd203f02eaca27daeb293&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR