Sodium-2-naphthalenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: 2-Naphthalenesulphonic acid Sodium Salt, 2-Naphthalenesulfonic acid Sodium Salt, Sodium 2-naphthalenesulphonate, Sodium 2-naphthalenesulfonate

N0016-500G N0016-25G
TCN0016-500GEA 439.71 CAD
TCN0016-500G TCN0016-025G
Sodium-2-naphthalenesulfonate ≥98.0%
Sodium-2-naphthalenesulfonate

CAS Number: 532-02-5
MDL Number: MFCD00064186
Molecular Formula: C10H8O3S
Molecular Weight: 230.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Naphthalenesulfonic Acid Sodium Salt

Formula: C₁₀H₈O₃S
MDL Number: MFCD00064186
CAS Number: 532-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed4bd8c337840d3867c93c6e463ed14a0d4d7f38&vid=ed4bd8c337840d3867c93c6e463ed14a0d4d7f38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR