Sodium 1-Naphthalenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Naphthalene-1-sulfonic acid sodium salt, 1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt, 1-Naphthalenesulfonic acid sodium salt

N0015-25G N0015-500G N0015-100G
TCN0015-025GEA 51.44 CAD
TCN0015-025G TCN0015-500G TCN0015-100G
Sodium 1-Naphthalenesulfonate ≥98.0%
Sodium 1-Naphthalenesulfonate

CAS Number: 130-14-3
MDL Number: MFCD00064964
Molecular Formula: C10H8O3S
Molecular Weight: 230.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1-Naphthalenesulfonic Acid Sodium Salt

Formula: C₁₀H₈O₃S
MDL Number: MFCD00064964
CAS Number: 130-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ef74d0df8f367b4f88930b85a0cfe1bae51d0d8&vid=4ef74d0df8f367b4f88930b85a0cfe1bae51d0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR