4-Nitro-m-anisic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxy-4-nitrobenzoic acid

M2083-25G M2083-5G
TCM2083-25GEA 219.62 CAD
TCM2083-25G TCM2083-5G
4-Nitro-m-anisic acid ≥98.0%
4-Nitro-m-anisic acid

CAS Number: 5081-36-7
MDL Number: MFCD00007353
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 235


Other Product Names / Synonyms: 4-Nitro-m-anisic Acid

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Boiling Pt: 405
Melting Pt: 233
MDL Number: MFCD00007353
CAS Number: 5081-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a6248e921ed1dd267936847386070be4612fac7&vid=c57ac0f931dbfad61f25f3ad593030df0f4f0442&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR