Methyl phenylpropiolate ≥98.0%

Supplier: TCI America

M2080-5G M2080-25G
TCM2080-5GEA 140.62 CAD
TCM2080-5G TCM2080-25G
Methyl phenylpropiolate ≥98.0%
Methyl phenylpropiolate

CAS Number: 4891-38-7
MDL Number: MFCD00041685
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 103
Freezing point (°C): 23


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Phenylpropiolic Acid Methyl Ester, Methyl Phenylpropargylate, Phenylpropargylic Acid Methyl Ester

Formula: C₁₀H₈O₂
Boiling Pt: 103 °C
MDL Number: MFCD00041685
CAS Number: 4891-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=179c0b035dfb3fa3b575e796d9de4957e0395074&vid=179c0b035dfb3fa3b575e796d9de4957e0395074&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR