3-(Methylthio)-1-hexanol ≥98.0%

Supplier: TCI America

Synonyms: 3-(Methylthio)hexan-1-ol, 3-(Methylthio)hexanol

M2050-5G M2050-25G
TCM2050-5GEA 58.46 CAD
TCM2050-5G TCM2050-25G
3-(Methylthio)-1-hexanol ≥98.0%
3-(Methylthio)-1-hexanol

CAS Number: 51755-66-9
MDL Number: MFCD00010674
Molecular Formula: C7H16OS
Molecular Weight: 148.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): 108
Specific Gravity (20/20): 0.97

Formula: C₄H₁₀OS
MW: 148.26 g/mol
Boiling Pt: 62 °C
Density: 0.97 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00010674
CAS Number: 51755-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a92d578fc1a231a7aa724d2c39c6d2679710d0bd&vid=a92d578fc1a231a7aa724d2c39c6d2679710d0bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR