Methyl indole-6-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 1H-indole-6-carboxylate

M2042-1G M2042-5G M2042-25G
TCM2042-1GEA 42.66 CAD
TCM2042-1G TCM2042-5G TCM2042-25G
Methyl indole-6-carboxylate ≥98.0%
Methyl indole-6-carboxylate

CAS Number: 50820-65-0
MDL Number: MFCD00211063
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77


Other Product Names / Synonyms: Indole-6-carboxylic Acid Methyl Ester

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00211063
CAS Number: 50820-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a2ff9672f9402f65cd1a225a08161e253b5336a&vid=2a2ff9672f9402f65cd1a225a08161e253b5336a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR