3,4-(Methylenedioxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3,4-(Methylenedioxy)benzeneboronic acid, Benzo[d][1,3]dioxol-5-ylboronic acid
M2035-25G
M2035-5G
TCM2035-25GEA
483.48
CAD
TCM2035-25G
TCM2035-5G
3,4-(Methylenedioxy)phenylboronic acid (contains varying amounts of Anhydride)
3,4-(Methylenedioxy)phenylboronic acid
CAS Number: 94839-07-3
MDL Number: MFCD01009695
Molecular Formula: C7H7BO4
Molecular Weight: 165.94
Form: Crystal
Color: White
Melting point (°C): 229
Other Product Names / Synonyms: 3,4-(Methylenedioxy)benzeneboronic Acid
Formula:
C₇H₇BO₄ MW: 165.94 g/mol Melting Pt: 229 °C |
MDL Number:
MFCD01009695 CAS Number: 94839-07-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...