Methyl-2-bromo-5-chlorobenzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-bromo-5-chlorobenzoate
M2033-25G
M2033-5G
TCM2033-25GEA
124.82
CAD
TCM2033-25G
TCM2033-5G
Methyl-2-bromo-5-chlorobenzoate ≥98.0%
Methyl 2-bromo-5-chlorobenzoate
CAS Number: 27007-53-0
MDL Number: MFCD00144763
Molecular Formula: C8H6BrClO2
Molecular Weight: 249.49
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41
Other Product Names / Synonyms: 2-Bromo-5-chlorobenzoic Acid Methyl Ester
Formula:
C₈H₆BrClO₂ Melting Pt: 41 °C |
MDL Number:
MFCD00144763 CAS Number: 27007-53-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...