Methyl-2-pyrazinecarboxylate ≥98.0%

Supplier: TCI America

M2015-5G M2015-25G
TCM2015-5GEA 139.04 CAD
TCM2015-5G TCM2015-25G
Methyl-2-pyrazinecarboxylate ≥98.0%
Methyl 2-pyrazinecarboxylate

CAS Number: 6164-79-0
MDL Number: MFCD00014611
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 104
Melting point (°C): 60


Other Product Names / Synonyms: 2-Pyrazinecarboxylic Acid Methyl Ester

Formula: C₆H₆N₂O₂
Boiling Pt: 104 °C
Melting Pt: 60 °C
MDL Number: MFCD00014611
CAS Number: 6164-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75ef9588dab32429b8a22fea5d379f7ae7c6589b&vid=a5eb3e54bdf62bbe4a11f51fd23238fabe5948fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR