Methyl trans-2-octenoate ≥95.0%
Supplier: TCI America
Synonyms:
trans-2-Octenoic Acid Methyl Ester, Methyl trans-2-Octenoate, (E)-2-Methyl oct-2-enoate
M2014-25ML
M2014-5ML
TCM2014-25MLEA
578.28
CAD
TCM2014-25ML
TCM2014-5ML
Methyl trans-2-octenoate ≥95.0%
Methyl trans-2-octenoate
CAS Number: 7367-81-9
MDL Number: MFCD00036620
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 83
Specific Gravity (20/20): 0.91
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: trans-2-Octenoic Acid Methyl Ester
Formula:
C₉H₁₆O₂ MW: 156.23 g/mol Boiling Pt: 91 °C Density: 0.91 (20 °C) Flash Pt: 83 °C |
MDL Number:
MFCD00036620 CAS Number: 7367-81-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...