Methyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2008-5G
M2008-1G
TCM2008-5GEA
133.83
CAD
TCM2008-5G
TCM2008-1G
Methyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)
Methyl 5-bromonicotinate
CAS Number: 29681-44-5
MDL Number: MFCD00173668
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 99
Other Product Names / Synonyms: 5-Bromo-3-pyridinecarboxylic Acid Methyl Ester, Methyl 5-Bromo-3-pyridinecarboxylate, 5-Bromonicotinic Acid Methyl Ester
Formula:
C₇H₆BrNO₂ Melting Pt: 99 °C |
MDL Number:
MFCD00173668 CAS Number: 29681-44-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...