Methyl-2,5-dihydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester

M2006-25G M2006-5G
TCM2006-25GEA 239.56 CAD
TCM2006-25G TCM2006-5G
Methyl-2,5-dihydroxybenzoate ≥98.0%
Methyl 2,5-dihydroxybenzoate

CAS Number: 2150-46-1
MDL Number: MFCD00016464
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 87


Other Product Names / Synonyms: 2,5-Dihydroxybenzoic Acid Methyl Ester, Methyl Gentisate, Gentisic Acid Methyl Ester

Formula: C₈H₈O₄
Boiling Pt: 136 °C
Melting Pt: 87 °C
MDL Number: MFCD00016464
CAS Number: 2150-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37453e90be2047c9dc57e833625da1b7f0e0d2e6&vid=37453e90be2047c9dc57e833625da1b7f0e0d2e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR