Methyl-4-amino-2-methoxybenzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 4-amino-2-methoxybenzenecarboxylate, 4-Amino-2-methoxybenzoic acid methyl ester, Methyl 4-amino-2-methoxybenzoate
M1998-25G
M1998-5G
TCM1998-25GEA
137.46
CAD
TCM1998-25G
TCM1998-5G
Methyl-4-amino-2-methoxybenzoate ≥98.0%
Methyl 4-amino-2-methoxybenzoate
CAS Number: 27492-84-8
MDL Number: MFCD00017202
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159
Other Product Names / Synonyms: 4-Amino-2-methoxybenzoic Acid Methyl Ester, Methyl 4-Amino-o-anisate, 4-Amino-o-anisic Acid Methyl Ester
Formula:
C₉H₁₁NO₃ Melting Pt: 159 °C |
MDL Number:
MFCD00017202 CAS Number: 27492-84-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...