Methyl-5-chloro-2-nitrobenzoate ≥98.0%
Supplier: TCI America
M1996-25G
M1996-5G
TCM1996-25GEA
42.66
CAD
TCM1996-25G
TCM1996-5G
Methyl-5-chloro-2-nitrobenzoate ≥98.0%
Methyl 5-chloro-2-nitrobenzoate
CAS Number: 51282-49-6
MDL Number: MFCD00272139
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50
Flash Point (°C): 110
Other Product Names / Synonyms: 5-Chloro-2-nitrobenzoic Acid Methyl Ester
Formula:
C₈H₆ClNO₄ Melting Pt: 50 °C Flash Pt: 110 °C |
MDL Number:
MFCD00272139 CAS Number: 51282-49-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...