Methyl-6-chloronicotinate ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

M1980-5G M1980-25G
TCM1980-5GEA 90.85 CAD
TCM1980-5G TCM1980-25G
Methyl-6-chloronicotinate ≥98.0% (by GC, total nitrogen)
Methyl 6-chloronicotinate

CAS Number: 73781-91-6
MDL Number: MFCD00023420
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 6-Chloronicotinic Acid Methyl Ester, Methyl 6-Chloro-3-pyridinecarboxylate, 6-Chloro-3-pyridinecarboxylic Acid Methyl Ester

Formula: C₇H₆ClNO₂
Melting Pt: 87 °C
MDL Number: MFCD00023420
CAS Number: 73781-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10f06c7f5aa61185548b58606ec8671757e1524b&vid=10f06c7f5aa61185548b58606ec8671757e1524b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR