4-Methoxybenzoic anhydride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M1973-5G M1973-25G
TCM1973-5GEA 251.22 CAD
TCM1973-5G TCM1973-25G
4-Methoxybenzoic anhydride ≥97.0% (by GC, titration analysis)
4-Methoxybenzoic anhydride

CAS Number: 794-94-5
MDL Number: MFCD00017175
Molecular Formula: C16H14O5
Molecular Weight: 286.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 98


Other Product Names / Synonyms: p-Anisic Anhydride, PMBA

Formula: C₁₆H₁₄O₅
Boiling Pt: 310 °C
Melting Pt: 98 °C
MDL Number: MFCD00017175
CAS Number: 794-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94c0616c3eea4fff4fd1150be1102b7919395b58&vid=94c0616c3eea4fff4fd1150be1102b7919395b58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR