4-(Methylsulfonyl)phenylboronic acid ≥98% (contains varying amounts of Anhydride)
Supplier: TCI America
M1972-1G
M1972-5G
TCM1972-1GEA
60.04
CAD
TCM1972-1G
TCM1972-5G
4-(Methylsulfonyl)phenylboronic acid ≥98% (contains varying amounts of Anhydride)
4-(Methylsulphonyl)phenylboronic acid
CAS Number: 149104-88-1
MDL Number: MFCD01630820
Molecular Formula: C7H9BO4S
Molecular Weight: 200.02
Form: Crystal
Color: White
Melting point (°C): 293
Other Product Names / Synonyms: 4-(Methanesulfonyl)phenylboronic Acid, 4-(Methanesulfonyl)benzeneboronic Acid
Formula:
C₇H₉BO₄S Melting Pt: 293 °C |
MDL Number:
MFCD01630820 CAS Number: 149104-88-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...