2-(Methylthio)benzoic acid ≥98.0%

Supplier: TCI America

M1953-25G M1953-5G
TCM1953-25GEA 436.08 CAD
TCM1953-25G TCM1953-5G
2-(Methylthio)benzoic acid ≥98.0%
2-(Methylthio)benzoic acid

CAS Number: 3724-10-5
MDL Number: MFCD00029934
Molecular Formula: C8H8O2S
Molecular Weight: 168.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 170

Formula: C₈H₈O₂S
MW: 168.22
Melting Pt: 170 °C
MDL Number: MFCD00029934
CAS Number: 3724-10-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85ed8584e26ca64e30247361335105586d29ef8a&vid=85ed8584e26ca64e30247361335105586d29ef8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR