3-Methylbenzo[b]thiophene ≥97.0%

Supplier: TCI America

M1952-5G M1952-1G
TCM1952-5GEA 325.48 CAD
TCM1952-5G TCM1952-1G
3-Methylbenzo[b]thiophene ≥97.0%
3-Methylbenzo[b]thiophene

CAS Number: 1455-18-1
MDL Number: MFCD00040243
Molecular Formula: C9H8S
Molecular Weight: 148.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -40
Flash Point (°C): 104
Specific Gravity (20/20): 1.13

Formula: C₉H₈S
Boiling Pt: 127 °C
Melting Pt: –40 °C
Density: 1.13 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00040243
CAS Number: 1455-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a88a972a86731d48d761039fb1d93b0e5bb0ea7&vid=2a88a972a86731d48d761039fb1d93b0e5bb0ea7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR