Methyl-3,4-dihydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate

M1943-25G M1943-5G
TCM1943-25GEA 153.26 CAD
TCM1943-25G TCM1943-5G
Methyl-3,4-dihydroxybenzoate ≥98.0%
Methyl 3,4-dihydroxybenzoate

CAS Number: 2150-43-8
MDL Number: MFCD00016437
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: 3,4-Dihydroxybenzoic Acid Methyl Ester, Methyl Protocatechuate, Protocatechuic Acid Methyl Ester

Formula: C₈H₈O₄
Melting Pt: 136 °C
MDL Number: MFCD00016437
CAS Number: 2150-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd0cb041201ecf2d41072ddf25d5463698d664e5&vid=fd0cb041201ecf2d41072ddf25d5463698d664e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR