(S)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1938-1G
M1938-5G
TCM1938-1GEA
196.71
CAD
TCM1938-1G
TCM1938-5G
(S)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC, titration analysis)
(S)-1-Methyl-3-pyrrolidinol
CAS Number: 104641-59-0
MDL Number: MFCD03788748
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 71
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: 6 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-3-Hydroxy-1-methylpyrrolidine
Formula:
C₅H₁₁NO Boiling Pt: 182 °C Density: 0.98 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD03788748 CAS Number: 104641-59-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...