Methyl-4-Chloro-3-methylbenzoate ≥98.0%

Supplier: TCI America

M1935-25G M1935-5G
TCM1935-25GEA 237.95 CAD
TCM1935-25G TCM1935-5G
Methyl-4-Chloro-3-methylbenzoate ≥98.0%
Methyl-4-Chloro-3-methylbenzoate

CAS Number: 91367-05-4
MDL Number: MFCD00151838
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 4-Chloro-3-methylbenzoic Acid Methyl Ester

Formula: C₉H₉ClO₂
Density: 1.21 (20 °C)
MDL Number: MFCD00151838
CAS Number: 91367-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f27f88197d5af8bb097d849a8a2d6d411f1d80d&vid=2f27f88197d5af8bb097d849a8a2d6d411f1d80d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR