Methyl-2-bromo-5-methoxybenzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-bromo-5-methoxybenzoate, 2-Bromo-5-methoxybenzoic acid methyl ester
M1934-25G
M1934-5G
TCM1934-25GEA
175.38
CAD
TCM1934-25G
TCM1934-5G
Methyl-2-bromo-5-methoxybenzoate ≥98.0%
Methyl 2-bromo-5-methoxybenzoate
CAS Number: 35450-36-3
MDL Number: MFCD00051594
Molecular Formula: C9H9BrO3
Molecular Weight: 245.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 110
Specific Gravity (20/20): 1.52
Other Product Names / Synonyms: 2-Bromo-5-methoxybenzoic Acid Methyl Ester, Methyl 6-Bromo-m-anisate, 6-Bromo-m-anisic Acid Methyl Ester
Formula:
C₉H₉BrO₃ Boiling Pt: 162 °C Density: 1.52 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00051594 CAS Number: 35450-36-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...