4-(Methylsulfonyl)benzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Methylsulphonylbenzoic acid, 4-Methylsulfonylbenzoic acid

M1932-5G M1932-25G
TCM1932-5GEA 56.88 CAD
TCM1932-5G TCM1932-25G
4-(Methylsulfonyl)benzoic acid ≥98.0%
4-(Methylsulphonyl)benzoic acid

CAS Number: 4052-30-6
MDL Number: MFCD00007564
Molecular Formula: C8H8O4S
Molecular Weight: 200.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 265

Formula: C₈H₈O₄S
MW: 200.21 g/mol
Melting Pt: 265 °C
MDL Number: MFCD00007564
CAS Number: 4052-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e95c59eba5ee8bb2a03f8487627b0c392a88589&vid=6e95c59eba5ee8bb2a03f8487627b0c392a88589&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR