Methyl-3,5-dibromobenzoate ≥98.0%

Supplier: TCI America

M1929-25G M1929-5G
TCM1929-25GEA 459.78 CAD
TCM1929-25G TCM1929-5G
Methyl-3,5-dibromobenzoate ≥98.0%
Methyl 3,5-dibromobenzoate

CAS Number: 51329-15-8
MDL Number: MFCD00082648
Molecular Formula: C8H6Br2O2
Molecular Weight: 293.94
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Dibromobenzoic Acid Methyl Ester

Formula: C₈H₆Br₂O₂
Melting Pt: 63 °C
MDL Number: MFCD00082648
CAS Number: 51329-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b7a26bae2270adbb7776d0497fc5ab205787d06&vid=6b7a26bae2270adbb7776d0497fc5ab205787d06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR