Methyl-4-fluoro-3-hydroxybenzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 4-fluoro-3-hydroxybenzoate
M1919-25G
M1919-5G
TCM1919-25GEA
589.34
CAD
TCM1919-25G
TCM1919-5G
Methyl-4-fluoro-3-hydroxybenzoate ≥98.0%
Methyl 4-fluoro-3-hydroxybenzoate
CAS Number: 214822-96-5
MDL Number: MFCD06797457
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-3-hydroxybenzoic Acid Methyl Ester
Formula:
C₈H₇FO₃ MW: 170.14 g/mol Melting Pt: 92 °C |
MDL Number:
MFCD06797457 CAS Number: 214822-96-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...