Methyl perfluorododecanoate ≥95.0%

Supplier: TCI America

M1917-25G M1917-5G
TCM1917-25GEA 259.44 CAD
TCM1917-25G TCM1917-5G
Methyl perfluorododecanoate ≥95.0%
Methyl perfluorododecanoate

CAS Number: 56554-52-0
MDL Number: MFCD00236616
Molecular Formula: C13H3F23O2
Molecular Weight: 628.13
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 58


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Methyl Perfluorododecanoate, Methyl Tricosafluorolaurate, Tricosafluorododecanoic Acid Methyl Ester, Perfluorododecanoic Acid Methyl Ester

Formula: C₁₃H₃F₂₃O₂
Boiling Pt: 125 °C
Melting Pt: 58 °C
MDL Number: MFCD00236616
CAS Number: 56554-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48e9665a74e5907d9abac0ca6e6080d5879d48d9&vid=48e9665a74e5907d9abac0ca6e6080d5879d48d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR