Methyl-4-iodsalicylate ≥98.0%

Supplier: TCI America

M1908-25G M1908-5G
TCM1908-25GEA 507.18 CAD
TCM1908-25G TCM1908-5G
Methyl-4-iodsalicylate ≥98.0%
Methyl 4-iodsalicylate

CAS Number: 18179-39-0
MDL Number: MFCD06797864
Molecular Formula: C8H7IO3
Molecular Weight: 278.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70


Other Product Names / Synonyms: Methyl 2-Hydroxy-4-iodobenzoate, 4-Iodosalicylic Acid Methyl Ester, 2-Hydroxy-4-iodobenzoic Acid Methyl Ester

Formula: C₈H₇IO₃
Melting Pt: 70 °C
MDL Number: MFCD06797864
CAS Number: 18179-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2bc8a6eefec79a8819862b2a27365469edfa8d1&vid=d2bc8a6eefec79a8819862b2a27365469edfa8d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR