4-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

M1907-5G M1907-25G M1907-1G
TCM1907-5GEA 58.46 CAD
TCM1907-5G TCM1907-25G TCM1907-1G
4-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Methoxycarbonyl)phenylboronic acid

CAS Number: 99768-12-4
MDL Number: MFCD01632203
Molecular Formula: C8H9BO4
Molecular Weight: 179.97
Form: Crystal
Color: White
Melting point (°C): 234


Other Product Names / Synonyms: 4-(Methoxycarbonyl)benzeneboronic Acid

Formula: C₈H₉BO₄
Melting Pt: 234 °C
MDL Number: MFCD01632203
CAS Number: 99768-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38404ded914a202ba8518c23fa56e6bcc7b17f66&vid=38404ded914a202ba8518c23fa56e6bcc7b17f66&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR