2-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

M1905-1G M1905-5G
TCM1905-1GEA 44.24 CAD
TCM1905-1G TCM1905-5G
2-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Methoxycarbonyl)phenylboronic acid

CAS Number: 374538-03-1
MDL Number: MFCD02179452
Molecular Formula: C8H9BO4
Molecular Weight: 179.97
Form: Crystal
Color: White
Melting point (°C): 104


Other Product Names / Synonyms: 2-(Methoxycarbonyl)benzeneboronic Acid

Formula: C₈H₉BO₄
Melting Pt: 104 °C
MDL Number: MFCD02179452
CAS Number: 374538-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73913639b1106e3e8a1ee7577e7139369e690d86&vid=73913639b1106e3e8a1ee7577e7139369e690d86&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR