Methyl-6-methylnicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 6-methylnicotinate

M1893-5G M1893-25G
TCM1893-5GEA 66.36 CAD
TCM1893-5G TCM1893-25G
Methyl-6-methylnicotinate ≥98.0% (by GC, titration analysis)
Methyl 6-methylnicotinate

CAS Number: 5470-70-2
MDL Number: MFCD00006340
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 34
Flash Point (°C): 103


Other Product Names / Synonyms: 6-Methylnicotinic Acid Methyl Ester, Methyl 6-Methylpyridine-3-carboxylate, 6-Methylpyridine-3-carboxylic Acid Methyl Ester

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 160 °C
Melting Pt: 34 °C
Flash Pt: 103 °C
MDL Number: MFCD00006340
CAS Number: 5470-70-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab7304c6db04e0fe1408b0741686614f6299845b&vid=ab7304c6db04e0fe1408b0741686614f6299845b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR