Methyl thiosalicylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-mercaptobenzoate

M1881-25G M1881-5G
TCM1881-25GEA 423.44 CAD
TCM1881-25G TCM1881-5G
Methyl thiosalicylate ≥98.0%
Methyl thiosalicylate

CAS Number: 4892-02-8
MDL Number: MFCD00060678
Molecular Formula: C8H8O2S
Molecular Weight: 168.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 113
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: Thiosalicylic Acid Methyl Ester, Methyl 2-Mercaptobenzoate, 2-Mercaptobenzoic Acid Methyl Ester

Formula: C₈H₈O₂S
MW: 168.21 g/mol
Boiling Pt: 123 °C
Density: 1.23 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00060678
CAS Number: 4892-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d16458bc0461bda8f11e9d283e65fa200c6c165&vid=0d16458bc0461bda8f11e9d283e65fa200c6c165&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR