Methyl-2-amino-5-chlorobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1877-5G M1877-1G
TCM1877-5GEA 109.02 CAD
TCM1877-5G TCM1877-1G
Methyl-2-amino-5-chlorobenzoate ≥98.0% (by GC, titration analysis)
Methyl 2-amino-5-chlorobenzoate

CAS Number: 5202-89-1
MDL Number: MFCD00007837
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 70


Other Product Names / Synonyms: 5-Chloroanthranilic Acid Methyl Ester, Methyl 2-Amino-5-chlorobenzoate, 2-Amino-5-chlorobenzoic Acid Methyl Ester

Formula: C₈H₈ClNO₂
Boiling Pt: 170 °C
Melting Pt: 70 °C
MDL Number: MFCD00007837
CAS Number: 5202-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd429b188dc6eea4d787d4df8054df385f8f4ff1&vid=bd429b188dc6eea4d787d4df8054df385f8f4ff1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR