Methyl-2-(aminosulfonyl)benzoate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2-(Aminosulfonyl)benzoic acid Methyl Ester, Methyl 2-sulphamoylbenzoate, 2-sulphamoylbenzoic acid Methyl Ester, Methyl 2-(Aminosulphonyl)benzoate, Methyl 2-sulfonamidobenzoate, Methyl 2-sulfamoylbenzoate, 2-(Aminosulphonyl)benzoic acid Methyl Ester, 2-sulphonamidobenzoic acid Methyl Ester, Methyl 2-sulphonamidobenzoate, Methyl 2-(Aminosulfonyl)benzoate, 2-sulfonamidobenzoic acid Methyl Ester, 2-sulfamoylbenzoic acid Methyl Ester

M1866-25G M1866-500G
TCM1866-25GEA 46.29 CAD
TCM1866-25G TCM1866-500G
Methyl-2-(aminosulfonyl)benzoate ≥98.0% (by total nitrogen basis)
Methyl-2-(aminosulfonyl)benzoate

CAS Number: 57683-71-3
MDL Number: MFCD00009808
Molecular Formula: C8H9NO4S
Molecular Weight: 215.22
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 2-(Aminosulfonyl)benzoic Acid Methyl Ester, Methyl 2-Sulfamoylbenzoate, 2-Sulfamoylbenzoic Acid Methyl Ester, Methyl 2-Sulfonamidobenzoate, 2-Sulfonamidobenzoic Acid Methyl Ester

Formula: C₈H₉NO₄S
MW: 215.22 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00009808
CAS Number: 57683-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92b0971f9d17431b829ceb6bbc7c49eb060a27c9&vid=92b0971f9d17431b829ceb6bbc7c49eb060a27c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR