3-(Methylthio)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1853-1G M1853-5G
TCM1853-1GEA 178.54 CAD
TCM1853-1G TCM1853-5G
3-(Methylthio)benzoic acid ≥98.0% (by GC, titration analysis)
3-(Methylthio)benzoic acid

CAS Number: 825-99-0
MDL Number: MFCD00156993
Molecular Formula: C8H8O2S
Molecular Weight: 168.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 128

Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00156993
CAS Number: 825-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cb9c02d07b1bf5f89636cb3c40709356d565f1b&vid=6cb9c02d07b1bf5f89636cb3c40709356d565f1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR