5-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
5-Methylthiophene-2-boronic acid, (5-Methylthiophen-2-yl)boronic acid
M1850-1G
M1850-5G
TCM1850-1GEA
64.46
CAD
TCM1850-1G
TCM1850-5G
5-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
5-Methyl-2-thiopheneboronic acid
CAS Number: 162607-20-7
MDL Number: MFCD01318166
Molecular Formula: C5H7BO2S
Molecular Weight: 141.98
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 150
Storage Temperature: <0°C
Formula:
C₅H₇BO₂S MW: 141.99 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD01318166 CAS Number: 162607-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...